MilliporeSigma
  • Toll-like receptor 4 signaling is involved in IgA-stimulated mesangial cell activation.
Toll-like receptor 4 signaling is involved in IgA-stimulated mesangial cell activation.
Yonsei medical journal (2011-05-31)
Beom Jin Lim, Dahye Lee, Soon Won Hong, Hyeon Joo Jeong
ABSTRACT
Deposition of polymeric IgA1 in the kidney mesangium is the hallmark of IgA nephropathy, but the molecular mechanisms of IgA-mediated mesangial responses and inflammatory injuries remain poorly understood. We hypothesize that Toll-like receptor 4 (TLR4) is involved in IgA-induced mesangial cell activation. Mouse mesangial cells were stimulated with lipopolysaccharide (LPS) (1 μg/mL), IgA (20 μg/mL), or both, and TLR4 expression was measured by real time RT-PCR and Western blot. Intracellular responses to LPS or IgA were assessed by Western blot for ERK1/2, JNK, p38 MAP kinases (MAPKs), Iκ-Bα degradation and fibronectin secretion. MCP-1 secretion was assessed by ELISA. Small interfering RNA (siRNA) of TLR4 was used to confirm that the effects were caused by TLR4 activity. LPS- or IgA-treatment upregulated the levels of TLR4 mRNA and protein in cultured MMC at 24 h. LPS and IgA induced rapid phosphorylation of MAPKs, but degradation of Iκ-Bα was observed only in LPS-treated MMC. LPS, but not IgA, induced increased secretion of MCP-1 and fibronectin at 24 h or 48 h. Combined LPS and IgA treatment did not cause additional increases in TLR4 mRNA and protein levels or Iκ-Bα degradation, and MCP-1 and fibronectin secretions were less than with LPS alone. LPS- or IgA-induced TLR4 protein levels and MAPK activation were inhibited by transfection with TLR4 siRNA. These results indicate that the activation of MAPKs and MCP-1 secretion are mediated by TLR4, at least in part, in IgA-treated mesangial cells. TLR4 is involved in mesangial cell injury by induction of pro-inflammatory cytokines in IgA nephropathy.
Sigma-Aldrich
IgA from human colostrum, reagent grade, buffered aqueous solution