MilliporeSigma
  • De novo synthesis of 2-substituted syn-1,3-diols via an iterative asymmetric hydration strategy.
De novo synthesis of 2-substituted syn-1,3-diols via an iterative asymmetric hydration strategy.
The Journal of organic chemistry (2006-09-26)
Md Moinuddin Ahmed, Matthew S Mortensen, George A O'Doherty
ABSTRACT
The enantioselective syntheses of several protected 4-substituted syn-3,5-dihydroxy carboxylic esters have been achieved from the corresponding achiral (E,E)- or (E,Z)-1,3-dienoates. The route relies upon an enantio- and regioselective Sharpless dihydroxylation and a palladium-catalyzed reduction to form gamma-substituted delta-hydroxy-1-enoates. The resulting delta-hydroxy-1-enoates are subsequently converted into benzylidene-protected 4-substituted syn-3,5-dihydroxy carboxylic esters in one step. The benzylidene-protected 3,5-dihydroxy carboxylic esters are produced in good overall yields (20-54%) and high enantiomeric excess (73-97% ee).
Sigma-Aldrich
AD-mix-β
Sigma-Aldrich
(DHQ)2PHAL, ≥95%
Sigma-Aldrich
AD-mix-α