MilliporeSigma
  • CCAAT-enhancer binding protein delta (C/EBPδ) attenuates tubular injury and tubulointerstitial fibrogenesis during chronic obstructive nephropathy.
CCAAT-enhancer binding protein delta (C/EBPδ) attenuates tubular injury and tubulointerstitial fibrogenesis during chronic obstructive nephropathy.
Laboratory investigation; a journal of technical methods and pathology (2013-11-20)
JanWillem Duitman, Keren S Borensztajn, Willem P C Pulskens, Jaklien C Leemans, Sandrine Florquin, C Arnold Spek
ABSTRACT
CCAAT-enhancer-binding protein delta (C/EBPδ) is a transcription factor mainly known for its role in inflammation and apoptosis/proliferation. Considering that these are key processes in renal fibrosis, we hypothesized that C/EBPδ would potentiate renal fibrosis. In line with this hypothesis, C/EBPδ has recently been suggested to regulate the fibrotic response during glomerulonephritis. Here we determined the importance of C/EBPδ in the development of renal tubulointerstitial fibrosis by subjecting 8- to 12-week-old C/EBPδ-deficient mice and age- and sex-matched wild-type controls to the unilateral ureteral obstruction model. Mice were killed at 1, 3, or 7 days post surgery, and renal tissues were obtained for RNA, protein, and immunohistochemical analysis. We show that C/EBPδ deficiency resulted in a more profound fibrotic response as evident from enhanced tubular injury, collagen deposition in the interstitial area, and higher expression of transforming growth factor-β. Moreover, we show that the increase in renal fibrosis in C/EBPδ-deficient mice does not depend on an altered proliferation/apoptosis balance or on a differential inflammatory response in the obstructed kidney. In conclusion, our study provides direct evidence that C/EBPδ is a novel mediator of renal fibrosis. Modulating C/EBPδ expression could consequently be a potential antifibrotic strategy in patients with chronic kidney disease.
Product Description
Roche
TriPure Isolation Reagent, solution (ready-to-use), pkg of 50 mL (11667157001), pkg of 200 mL (11667165001)