MilliporeSigma
  • Role of the RhoA/ROCK pathway in high-altitude associated neonatal pulmonary hypertension in lambs.
Role of the RhoA/ROCK pathway in high-altitude associated neonatal pulmonary hypertension in lambs.
American journal of physiology. Regulatory, integrative and comparative physiology (2016-02-26)
Nandy C Lopez, German Ebensperger, Emilio A Herrera, Roberto V Reyes, Gloria Calaf, Gertrudis Cabello, Fernando A Moraga, Felipe A Beñaldo, Marcela Diaz, Julian T Parer, Anibal J Llanos
ABSTRACT
Exposure to high-altitude chronic hypoxia during pregnancy may cause pulmonary hypertension in neonates, as a result of vasoconstriction and vascular remodeling. We hypothesized that susceptibility to pulmonary hypertension, due to an augmented expression and activity of the RhoA/Rho-kinase (ROCK) pathway in these neonates, can be reduced by daily administration of fasudil, a ROCK inhibitor. We studied 10 highland newborn lambs with conception, gestation, and birth at 3,600 m in Putre, Chile. Five highland controls (HLC) were compared with 5 highland lambs treated with fasudil (HL-FAS; 3 mg·kg(-1)·day(-1) iv for 10 days). Ten lowland controls were studied in Lluta (50 m; LLC). During the 10 days of fasudil daily administration, the drug decreased pulmonary arterial pressure (PAP) and resistance (PVR), basally and during a superimposed episode of acute hypoxia. HL-FAS small pulmonary arteries showed diminished muscular area and a reduced contractile response to the thromboxane analog U46619 compared with HLC. Hypoxia, but not fasudil, changed the protein expression pattern of the RhoA/ROCKII pathway. Moreover, HL-FAS lungs expressed less pMYPT1(T850) and pMYPT1T(696) than HLC, with a potential increase of the myosin light chain phosphatase activity. Finally, hypoxia induced RhoA, ROCKII, and PKG mRNA expression in PASMCs of HLC, but fasudil reduced them (HL-FAS) similarly to LLC. We conclude that fasudil decreases the function of the RhoA/ROCK pathway, reducing the PAP and PVR in chronically hypoxic highland neonatal lambs. The inhibition of ROCKs by fasudil may offer a possible therapeutic tool for the pulmonary hypertension of the neonates.
Sigma-Aldrich
Anti-phospho-MYPT1 (Thr850) Antibody, Upstate®, from rabbit