MilliporeSigma
  • Impact of Organic Cation Transporters (OCT-SLC22A) on Differential Diagnosis of Intrahepatic Lesions.
Impact of Organic Cation Transporters (OCT-SLC22A) on Differential Diagnosis of Intrahepatic Lesions.
Drug metabolism and disposition: the biological fate of chemicals (2016-12-03)
Michele Visentin, Belle V van Rosmalen, Christian Hiller, Matthanja Bieze, Lia Hofstetter, Joanne Verheij, Gerd A Kullak-Ublick, Hermann Koepsell, Saffire S K S Phoa, Ikumi Tamai, Roelof J Bennink, Thomas M van Gulik, Bruno Stieger
ABSTRACT
Positron emission tomography (PET) using the cationic compound [
Product Description
Supelco
1-Methylpyridinium iodide, analytical standard