MilliporeSigma
Cell lineage responses to photobiomodulation therapy.
Journal of biophotonics (2016-07-09)
Karl W Engel, Imran Khan, Praveen R Arany
ABSTRACT
Photobiomodulation (PBM) therapy has been noted to promote cell proliferation and growth in many different cell types shown both in vitro and in vivo. Currently, treatment regimens are used in the clinic for a variety of ailments, including wound healing. However, most protocols treat an anatomical site without considering individual cell types constituting the target tissues. This study investigates the maximal dose threshold for oral keratinocyte and fibroblast cell types treated with near-infrared laser therapy. We observed keratinocytes have increased sensitivity to laser irradiances (>0.047 W/cm
Sigma-Aldrich
Glutathione Reductase Assay Kit, Sufficient for 100 colorimetric tests