MilliporeSigma
  • Excitation power dependent population pathways and absolute quantum yields of upconversion nanoparticles in different solvents.
Excitation power dependent population pathways and absolute quantum yields of upconversion nanoparticles in different solvents.
Nanoscale (2017-03-16)
C Würth, M Kaiser, S Wilhelm, B Grauel, T Hirsch, U Resch-Genger
ABSTRACT
The rational design of brighter upconversion nanoparticles (UCNPs) requires a better understanding of the radiationless deactivation pathways in these materials. Here, we demonstrate the potential of excitation power density (P)-dependent studies of upconversion (UC) luminescence intensities, slope factors, and absolute quantum yields (Φ
Sigma-Aldrich
Nitrosyl tetrafluoroborate, 95%