MilliporeSigma
  • Thermodynamics of enzyme-catalyzed esterifications: I. Succinic acid esterification with ethanol.
Thermodynamics of enzyme-catalyzed esterifications: I. Succinic acid esterification with ethanol.
Applied microbiology and biotechnology (2017-05-14)
Emrah Altuntepe, Thorsten Greinert, Felix Hartmann, Annika Reinhardt, Gabriele Sadowski, Christoph Held
ABSTRACT
Succinic acid (SA) was esterified with ethanol using Candida antarctica lipase B immobilized on acrylic resin at 40 and 50 °C. Enzyme activity in the reaction medium was assured prior to reaction experiments. Reaction-equilibrium experiments were performed for varying initial molalities of SA and water in the reaction mixtures. This allowed calculating the molality-based apparent equilibrium constant K
Sigma-Aldrich
mono-Ethyl succinate, 90%