MilliporeSigma
  • Lentivector Iterations and Pre-Clinical Scale-Up/Toxicity Testing: Targeting Mobilized CD34
Lentivector Iterations and Pre-Clinical Scale-Up/Toxicity Testing: Targeting Mobilized CD34
Molecular therapy. Methods & clinical development (2017-06-13)
Ju Huang, Aneal Khan, Bryan C Au, Dwayne L Barber, Lucía López-Vásquez, Nicole L Prokopishyn, Michel Boutin, Michael Rothe, Jack W Rip, Mona Abaoui, Murtaza S Nagree, Shaalee Dworski, Axel Schambach, Armand Keating, Michael L West, John Klassen, Patricia V Turner, Sandra Sirrs, C Anthony Rupar, Christiane Auray-Blais, Ronan Foley, Jeffrey A Medin
ABSTRACT
Fabry disease is a rare lysosomal storage disorder (LSD). We designed multiple recombinant lentivirus vectors (LVs) and tested their ability to engineer expression of human α-galactosidase A (α-gal A) in transduced Fabry patient CD34
Sigma-Aldrich
N-Acetyl-D-galactosamine, ≥98% (HPLC)