MilliporeSigma
  • Seaweed Extracts Enhance Salam Turfgrass Performance during Prolonged Irrigation Intervals and Saline Shock.
Seaweed Extracts Enhance Salam Turfgrass Performance during Prolonged Irrigation Intervals and Saline Shock.
Frontiers in plant science (2017-07-01)
Hosam O Elansary, Kowiyou Yessoufou, Amal M E Abdel-Hamid, Mohamed A El-Esawi, Hayssam M Ali, Mohamed S Elshikh
ABSTRACT
The negative effects of the ongoing climate change include unusual prolonged droughts and increased salinity pressures on the agricultural lands. Consequently, crops are facing unprecedented environmental pressure, and this calls for more research toward controlling such major stresses. The current study investigates the effects of seaweed extract sprays of
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests