MilliporeSigma
  • An examination of the effect of aerosolized Permanone insecticide on zebra finch susceptibility to West Nile virus.
An examination of the effect of aerosolized Permanone insecticide on zebra finch susceptibility to West Nile virus.
Environmental toxicology and chemistry (2017-07-20)
Mark D Jankowski, Murray E Moore, Erik K Hofmeister
ABSTRACT
West Nile virus (WNV) is maintained cryptically primarily in avian (passerine) populations, where it is transmitted by Culex spp. mosquitoes. Mosquito-control measures currently include physical activities to reduce mosquito-breeding sites and the application of mosquito larvicides or aerosolized insecticides to kill adults (adulticides) when arboviral diseases such as WNV or Zika virus are detected in mosquito populations. Organochlorine, organophosphorus, carbamate, and pyrethroid insecticides are often used. Previous work suggests an effect of pyrethroids on the immune system in a variety of vertebrates. We examined the effects of exposure to aerosolized Permanone® 30:30 insecticide (permethrin and piperonyl butoxide in soy oil vehicle) at approximately 10
Product Description
Supelco
Resmethrin, PESTANAL®, analytical standard, mixture of isomers