MilliporeSigma
  • Nitrifier-induced denitrification is an important source of soil nitrous oxide and can be inhibited by a nitrification inhibitor 3,4-dimethylpyrazole phosphate.
Nitrifier-induced denitrification is an important source of soil nitrous oxide and can be inhibited by a nitrification inhibitor 3,4-dimethylpyrazole phosphate.
Environmental microbiology (2017-07-29)
Xiuzhen Shi, Hang-Wei Hu, Xia Zhu-Barker, Helen Hayden, Juntao Wang, Helen Suter, Deli Chen, Ji-Zheng He
ABSTRACT
Soil ecosystem represents the largest contributor to global nitrous oxide (N
Sigma-Aldrich
Nitrosobenzene, ≥97%