- Melatonin reduces apoptotic cells, SOD2 and HSPB1 and improves the in vitro production and quality of bovine blastocysts.
Melatonin reduces apoptotic cells, SOD2 and HSPB1 and improves the in vitro production and quality of bovine blastocysts.
Reproduction in domestic animals = Zuchthygiene (2017-12-06)
T C Marques, E C da Silva Santos, T O Diesel, L O Leme, C F Martins, Man Dode, B G Alves, Fph Costa, E B de Oliveira, M L Gambarini
PMID29205523
ABSTRACT
Effects of adding different concentrations of melatonin (10