
8-Prenylnaringenin

Synonym(s):
2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-8-(3-methyl-2-butenyl)-4H-1-benzopyran-4-one
Empirical Formula (Hill Notation):
C20H20O5
CAS Number:
Molecular Weight:
340.37
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing