
BCIP

Synonym(s):
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt, BCIP® p-toluidine salt, X-phosphate p-toluidine salt
Empirical Formula (Hill Notation):
C8H6BrClNO4P · C7H9N
CAS Number:
Molecular Weight:
433.62
Products
Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing