
N-Sulfocarbamoylgonyautoxin-2-15N7 and N-Sulfocarbamoylgonyautoxin-3-15N7 solution

Synonym(s):
Toxin C1,2
Empirical Formula (Hill Notation):
C10H1715N7O11S2
Molecular Weight:
482.37
Products
Product Category
Brand
Formula Weight
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing