Skip to Content
Merck
Prednisone British Pharmacopoeia (BP) Reference Standard

Prednisone

Prednisone British Pharmacopoeia (BP) Reference Standard
Synonym(s):
1,4-Pregnadiene-17α,21-diol-3,11,20-trione, 1-Cortisone, 17α,21-Dihydroxy-1,4-pregnadiene-3,11,20-trione, Dehydrocortisone
Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility
Technique