Skip to Content
Merck
Z-Pro-OH 99%

Z-Pro-OH

Z-Pro-OH 99%
Synonym(s):
Z-L-Proline
Empirical Formula (Hill Notation):
C13H15NO4
CAS Number:
Molecular Weight:
249.26
EC Number:

Products

Shipping
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing