Merck
(+)-Catechin prevents human plasma oxidation.
Free radical biology & medicine (1998-01-23)
S B Lotito, C G Fraga
ABSTRACT
Based on the recognized capacity of (+)-catechin (CTCH) to prevent free radical-mediated damage in different biological systems, its role in the protection of human plasma from oxidation was investigated. Samples of human blood plasma were incubated with 50 mM AAPH [2,2'-azobis-(2-amidinopropane) clorhidrate] or AMVN [2,2'-azobis(2,4-valeronitrile)], in the absence or the presence of CTCH (0.01 to 1 mM). Lipid oxidation was evaluated measuring the formation of 2-thiobarbituric acid reactive substances (TBARS). Alpha-tocopherol (AT), beta-carotene (BC), and CTCH were measured by reverse phase HPLC with electrochemical detection. TBARS formation was dependent on incubation time and on the nature of the azocompound, yielding 4.8 +/- 0.9, and 14.9 +/- 3.4 microM MDA, after 4 h, in AAPH and AMVN-exposed plasma, respectively. Plasma AT and BC were extensively depleted under these oxidant conditions. The addition of CTCH prevented or delayed the formation of TBARS, and the depletion of AT and BC in a dose dependent manner. This antioxidant effect was dependent on the concentration of CTCH and on the physical characteristics of the radical initiator. CTCH supplementation modified not only the lag time for the antioxidants depletion, but also the consumption rate. These results indicate that CTCH was an effective antioxidant in human blood plasma, delaying the consumption of endogenous lipid soluble antioxidants (AT and BC) and inhibiting lipid oxidation.
Sigma-Aldrich
(+)-Catechin hydrate, ≥98% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
1 Available to ship on 16 December 2024
Details...
€107.00
10 g
1 Available to ship on 16 December 2024
Details...
€125.00