Merck
  • Modulation of a membrane lipid lamellar-nonlamellar phase transition by cationic lipids: a measure for transfection efficiency.
Modulation of a membrane lipid lamellar-nonlamellar phase transition by cationic lipids: a measure for transfection efficiency.
Biochimica et biophysica acta (2008-08-30)
Boris G Tenchov, Li Wang, Rumiana Koynova, Robert C MacDonald
ABSTRACT
Synthetic cationic lipids can be used as DNA carriers and are regarded to be the most promising non-viral gene carriers. For this investigation, six novel phosphatidylcholine (PC) cationic derivatives with various hydrophobic moieties were synthesized and their transfection efficiencies for human umbilical artery endothelial cells (HUAEC) were determined. Three compounds with relatively short, myristoleoyl or myristelaidoyl 14:1 chains exhibited very high activity, exceeding by approximately 10 times that of the reference cationic derivative dioleoyl ethylPC (EDOPC). Noteworthy, cationic lipids with 14:1 hydrocarbon chains have not been tested as DNA carriers in transfection assays previously. The other three lipids, which contained oleoyl 18:1 and longer chains, exhibited moderate to weak transfection activity. Transfection efficiency was found to correlate strongly with the effect of the cationic lipids on the lamellar-to-inverted hexagonal, Lalpha-->HII, phase conversion in dipalmitoleoyl phosphatidylethanolamine dispersions (DPoPE). X-ray diffraction on binary DPoPE/cationic lipid mixtures showed that the superior transfection agents eliminated the direct Lalpha-->HII phase transition and promoted formation of an inverted cubic phase between the Lalpha and HII phases. In contrast, moderate and weak transfection agents retained the direct Lalpha-->HII transition but shifted to higher temperatures than that of pure DPoPE, and induced cubic phase formation at a later stage. On the basis of current models of lipid membrane fusion, promotion of a cubic phase by the high-efficiency agents may be considered as an indication that their high transfection activity results from enhanced lipoplex fusion with cellular membranes. The distinct, well-expressed correlation established between transfection efficiency of a cationic lipid and the way it modulates nonlamellar phase formation of a membrane lipid could be useful as a criterion to assess the quality of lipid carriers and for rational design of new and superior nucleotide delivery agents.
Product Description
Avanti
14:1 EPC (Tf Salt), Avanti Research - A Croda Brand 890717O
Avanti
18:0 EPC (Cl Salt), Avanti Research - A Croda Brand 890703P, powder
Avanti
12:0 EPC (Cl Salt), Avanti Research - A Croda Brand 890700P, powder
Avanti
14:0 EPC (Cl Salt), Avanti Research - A Croda Brand
Avanti
18:0 EPC (Cl Salt), Avanti Research - A Croda Brand 890703C
Avanti
18:1 EPC (Cl Salt), Avanti Research - A Croda Brand 890704C
Avanti
18:1 EPC (Cl Salt), Avanti Research - A Croda Brand
Avanti
16:0-18:1 EPC (Cl Salt), Avanti Research - A Croda Brand 890705C
Avanti
16:0 EPC (Cl Salt), Avanti Research - A Croda Brand 890702P, powder
Avanti
16:0-18:1 EPC (Cl Salt), Avanti Research - A Croda Brand 890705P, powder