Merck
  • Protease-activated receptor 2 (PAR-2) antagonist AZ3451 as a novel therapeutic agent for osteoarthritis.
Protease-activated receptor 2 (PAR-2) antagonist AZ3451 as a novel therapeutic agent for osteoarthritis.
Aging (2019-12-17)
Xiaojian Huang, Bowei Ni, Yang Xi, Xiangyu Chu, Rui Zhang, Hongbo You
ABSTRACT
Osteoarthritis (OA) is a highly prevalent joint disorder blamed for pain and disability in older individuals. It's commonly accepted that inflammation, apoptosis, autophagy and cellular senescence participate in the progress of OA. Protease activated receptor 2 (PAR2), a member of the G-protein coupled receptors, is involved in the regulation of various inflammation diseases. Previous studies have identified PAR2 as a potential therapeutic target for the treatment of OA. Here, we investigated the role of PAR2 antagonist AZ3451 in inflammation response, apoptosis, autophagy and cellular senescence during OA. We confirmed that PAR2 expression was significantly up-regulated in OA articular cartilage tissues as well as in interleukin 1β (IL-1β) stimulated chondrocytes. We demonstrated AZ3451 could prevent the IL-1β-induced inflammation response, cartilage degradation and premature senescence in chondrocytes. Further study showed that AZ3451 attenuated chondrocytes apoptosis by activating autophagy in vitro. The P38/MAPK, NF-κB and PI3K/AKT/mTOR pathways were involved in the protective effect of AZ3451. In vivo, we found that intra-articular injection of AZ3451 could ameliorate the surgery induced cartilage degradation in rat OA model. Our work provided a better understanding of the mechanism of PAR2 in OA, and indicated that PAR2 antagonist AZ3451 might serve as a promising strategy for OA treatment.
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
AZ3451, ≥98% (HPLC)