Merck
  • The Role of Formamidine Groups in Dextran Based Nonviral Vectors for Gene Delivery on Their Physicochemical and Biological Characteristics.
The Role of Formamidine Groups in Dextran Based Nonviral Vectors for Gene Delivery on Their Physicochemical and Biological Characteristics.
Macromolecular bioscience (2020-10-08)
Dagmar Fischer, Niels Dusek, Konrad Hotzel, Thomas Heinze
ABSTRACT
Dextran-formamidine esters (dextran-N-[(dimethylamino)methylene]-β-alanine ester) with different degrees of substitution (0.45-0.92) are synthesized in an one-pot reaction. Dextran (Mw 60 000 g mol-1 ) is allowed to react with unprotected beta-alanine and iminium chloride and investigated regarding the potential as gene delivery system for the transfer of plasmid DNA. With degrees of substitution ≥ 0.63 improved DNA binding with formation of enzymatically stable complexes of about 130-160 nm with negative surface charges are obtained. These physicochemical characteristics correlated with increasing transfection rates in CHO-K1 cells determined by a luciferase reporter gene assay in dependency of the number of formamidine residues, N/P ratios and amount of DNA. The role of the number of formamidine groups is also highlighted by in vitro cyto- and hemotoxicity tests under the chosen conditions. These results indicate that dextran-formamidine esters are a very promising material for the safe and efficient gene delivery.
Sigma-Aldrich
sodium camptothecin, AldrichCPR
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000