Merck
  • Stress-Induced Metabolic Disorder in Peripheral CD4+ T Cells Leads to Anxiety-like Behavior.
Stress-Induced Metabolic Disorder in Peripheral CD4+ T Cells Leads to Anxiety-like Behavior.
Cell (2019-11-02)
Ke-Qi Fan, Yi-Yuan Li, Hao-Li Wang, Xin-Tao Mao, Jin-Xin Guo, Fei Wang, Ling-Jie Huang, Yi-Ning Li, Xiang-Yu Ma, Zheng-Jun Gao, Wei Chen, Dan-Dan Qian, Wen-Jin Xue, Qian Cao, Lei Zhang, Li Shen, Long Zhang, Chao Tong, Jiang-Yan Zhong, Wei Lu, Ling Lu, Ke-Ming Ren, Guisheng Zhong, Yuan Wang, Mingliang Tang, Xin-Hua Feng, Ren-Jie Chai, Jin Jin
ABSTRACT
Physical or mental stress leads to neuroplasticity in the brain and increases the risk of depression and anxiety. Stress exposure causes the dysfunction of peripheral T lymphocytes. However, the pathological role and underlying regulatory mechanism of peripheral T lymphocytes in mood disorders have not been well established. Here, we show that the lack of CD4+ T cells protects mice from stress-induced anxiety-like behavior. Physical stress-induced leukotriene B4 triggers severe mitochondrial fission in CD4+ T cells, which further leads to a variety of behavioral abnormalities including anxiety, depression, and social disorders. Metabolomic profiles and single-cell transcriptome reveal that CD4+ T cell-derived xanthine acts on oligodendrocytes in the left amygdala via adenosine receptor A1. Mitochondrial fission promotes the de novo synthesis of purine via interferon regulatory factor 1 accumulation in CD4+ T cells. Our study implicates a critical link between a purine metabolic disorder in CD4+ T cells and stress-driven anxiety-like behavior.
Sigma-Aldrich
Forodesine hydrochloride, ≥98% (HPLC)