Merck
Purification and properties of collagenase from a Streptomyces species.
Journal of biochemistry (1987-07-01)
A Endo, S Murakawa, H Shimizu, Y Shiraishi
ABSTRACT
A collagenase active against native, insoluble collagen was isolated from the culture filtrate of a streptomycete which has been designated Streptomyces sp. C-51. Collagenase was produced by growing this strain in media containing gelatin. Purification by ammonium sulfate fractionation and chromatographies on DEAE-Toyopearl and DEAE-Cellulofine columns produced active enzyme which was free of contaminating proteins, including nonspecific proteinases, but which contained two active subspecies (I and II). Both subspecies were purified by preparative slab gel electrophoresis. The apparent molecular weights were 100,000 for the homogeneous fraction I and 90,000-110,000 for the microheterogeneous fraction II. These two subspecies were most active at pH 8-9, very similar in amino acid composition and immunologically identical. Some other properties of the Streptomyces C-51 collagenase were compared with those of Clostridium histolyticum collagenase. Substrate specificity, insensitivity to N-ethylmaleimide and diisopropyl fluorophosphate, and sensitivity to certain metal ion complexing agents were similar for the collagenases from both microorganisms.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, Type V-S, ≥1 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type F, ≥2.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, suitable for cell culture, ≥4 FALGPA units/mg solid, high purity, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, purified by chromatography, ≥500 CDU/mg solid (CDU = collagen digestion units), lyophilized powder
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat epididymal adipocytes, Type II-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2μm filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat hepatocytes, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, non-sterile; 0.2 μm filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2 μm filtered solution), suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid