Merck
  • Ginsenosides from Panax ginseng differentially regulate lymphocyte proliferation.
Ginsenosides from Panax ginseng differentially regulate lymphocyte proliferation.
Planta medica (2002-07-03)
Jae Youl Cho, Ae Ra Kim, Eun Sook Yoo, Kyong Up Baik, Myung Hwan Park
ABSTRACT
We have examined the immunosuppressive effects of representative ginsenosides (Rb1, Rb2, Re and Rg1) from Panax ginseng C. A. Meyer on CD4+ and CD8+ lymphocyte proliferation. Ginsenosides differentially modulated lymphocyte proliferation induced by concanavalin A (Con A), lipopolysaccharide (LPS), phytohemaglutinin (PHA) and interleukin-2 (IL-2). Thus, Rb1 and Re significantly enhanced Con A-induced lymphocyte proliferation, whereas Rg1 did not affect the proliferation. Interestingly, however, Rb2 strongly blocked Con A, LPS and PHA-induced lymphocyte proliferation with the IC50 values of 21.8, 29.0 and 24.0 microM, respectively. Moreover, Rb2 inhibited Con A-stimulated IL-2 production with an IC 50 of 13.3 microM. In the IL-2-stimulated CD8+ T cell (CTLL-2) proliferation assay, Re and Rg1 showed strong suppressive effects with IC50 values of 57.5 and 64.7 microM, respectively. In contrast, neither Rb1 nor Rb2 did inhibit CTLL-2 cell proliferation at tested concentrations. These results suggest that ginsenosides from P. ginseng may modulate lymphocyte proliferation in a different manner.
Product Description
Supelco
Ginsenoside Rb2, analytical standard