Merck
  • Enantiomeric resolution of bifonazole by supercritical fluid chromatography.
Enantiomeric resolution of bifonazole by supercritical fluid chromatography.
Journal of separation science (2006-08-10)
Laura Toribio, Ma Jesús del Nozal, José L Bernal, Cristina Alonso, Juan J Jiménez
ABSTRACT
The enantiomeric separation of bifonazole by supercritical fluid chromatography on Chiralpak AD has been studied. The effect of different modifiers (methanol, ethanol, 2-propanol, and acetonitrile) was examined. Enantioseparation was possible with all of them, but the best results were provided by the alcohol-type ones. The resolution was higher than 5 in all cases. The isoelution temperatures Tiso were calculated from the study of the temperature effect for the different organic modifiers. The value of Tiso was below the working temperature range on using methanol, but above it with ethanol or 2-propanol.
Product Description
Bifonazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bifonazole, ≥98% (HPLC)
Supelco
Bifonazole, Pharmaceutical Secondary Standard; Certified Reference Material