Merck
  • Reversible fluorescence quenching by micelle selective benzophenone-induced interactions between brij micelles and polyacrylic acids: implications for chemical sensors.
Reversible fluorescence quenching by micelle selective benzophenone-induced interactions between brij micelles and polyacrylic acids: implications for chemical sensors.
The journal of physical chemistry. B (2010-08-19)
Prasun Bandyopadhyay, Amit K Ghosh
ABSTRACT
The fluorescence response of pyrene has been studied in the presence of nonionic brij micelles and poly(acrylic acid) (PAA) with benzophenone (BP) as a neutral hydrophobic quencher. Pyrene emission is quenched ("off" state) in the presence of BP in brij 35 (polyoxyethylene-23-lauryl ether) and brij 56 (polyoxyethylene-10-cetyl ether) micelles. Quenched pyrene emission is selectively recovered ("on" state) for brij 35 micelles with the addition of PAA (starting conc 2.0 x 10(-5) M). Due to the interaction of PAA and brij 35 micelles and the relatively easier accessibility of PAA polymer chains near the bulky polyoxyethylene chain of brij 35 micelles, the chances of BP partition inside the hydrophobic polymer coil are more compared to brij 56 micelles. The PAA sensing ability of the "brij 35:pyrene:BP" system is dependent on the molecular weight (M) of the polymer. Fluorescence recovery has been observed with PAA (M approximately 150000) and complete recovery has been recorded with high M of PAA (M approximately 450000); however, no fluorescence change is observed in the presence of low M of PAA (M approximately 2000). In solution, such selective reversible fluorescence quenching has the potential for a new class of highly sensitive chemical sensor systems.
Sigma-Aldrich
Brij® C10, average Mn ~683
Sigma-Aldrich
SP Brij® C2 MBAL-SO-(SG), average Mn ~330
Sigma-Aldrich
Brij® 58, average Mn ~1124
Sigma-Aldrich
ECO BRIJ® C10