Merck
Origin and nature of bond rotation barriers: a unified view.
The journal of physical chemistry. A (2013-01-19)
Shubin Liu
ABSTRACT
Bond rotations are common and important in molecular studies such as drug design and enzymatic reactions. Steric hindrance and hyperconjugation are often borrowed to justify the origin of bond rotation barriers. However, there exists no consensus, even for as simple molecules as ethane, on which the effect is more significant. Here, we show that a unified view is possible. To that end, we employ two energy partition schemes from density functional theory to investigate the flexible rotation barrier of six molecules with one rotatable dihedral angle: ethane, methylamine, methanol, hydrazine, hydroxylamine, and hydrogen peroxide. Our results suggest that, even though steric and quantum effects play indispensable roles, it is the electrostatic interaction that governs the barrier height of all these different types of systems. This work not only consolidates earlier views about the role of steric and quantum effects, it also provides new insights about the origin and nature of the bond rotation barrier, which should be applicable to many different types of chemical bonds.
Sigma-Aldrich
Hydrazine sulfate salt, ACS reagent, ≥99.0%
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Ethane, ≥99%
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine-13C hydrochloride, 99 atom % 13C
Sigma-Aldrich
Hydrazine dihydrochloride, ≥98%
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Hydrazine sulfate salt, puriss. p.a., ACS reagent, ≥99.0%
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Hydrazine solution, 1.0 M in THF
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Methylamine-15N hydrochloride, 98 atom % 15N
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
Sigma-Aldrich
Hydrazine hemisulfate salt, ≥98%
Sigma-Aldrich
Methylamine hydrochloride, ≥98%
Sigma-Aldrich
Hydrazine solution, 35 wt. % in H2O
Sigma-Aldrich
Hydrazine solution, 1 M in acetonitrile
Sigma-Aldrich
Hydrazine solution, 1.0 M in ethanol