Merck
  • Localization of the insertion site and pathotype determination of the locus of enterocyte effacement of shiga toxin-producing Escherichia coli strains.
Localization of the insertion site and pathotype determination of the locus of enterocyte effacement of shiga toxin-producing Escherichia coli strains.
Applied and environmental microbiology (2004-01-09)
Yolande Bertin, Karima Boukhors, Valerie Livrelli, Christine Martin
ABSTRACT
Of 220 Shiga toxin-producing Escherichia coli (STEC) strains collected in central France from healthy cattle, food samples, and asymptomatic children, 12 possessed the eae gene included in the locus of enterocyte effacement (LEE) pathogenicity island. Based on gene typing, we observed 7 different eae espA espB tir pathotypes among the 12 STEC strains and described the new espAbetav variant. As previously observed, the O157 serogroup is associated with eaegamma, O26 is associated with eaebeta, and O103 is associated with eaeepsilon. However, the unexpected eaezeta allele was detected in 5 of the 12 isolates. PCR amplification and pulsed-field gel electrophoresis using the I-CeuI endonuclease followed by Southern hybridization indicated that the LEE was inserted in the vicinity of the selC (three isolates), pheU (two isolates), or pheV (six isolates) tRNA gene. Six isolates harbored two or three of these tRNA loci altered by the insertion of integrase genes (CP4-int and/or int-phe), suggesting the insertion of additional foreign DNA fragments at these sites. In spite of great genetic diversity of LEE pathotypes and LEE insertion sites, bovine strains harbor alleles of LEE genes that are frequently found in clinical STEC strains isolated from outbreaks and sporadic cases around the world, underscoring the potential risk of the bovine strains on human health.
Sigma-Aldrich
AccuTaq LA 10× Buffer, 10X Buffer for long and accurate PCR