Merck
Fatal methemoglobinemia due to inhalation of isobutyl nitrite.
Journal of toxicology. Clinical toxicology (1994-01-01)
S M Bradberry, R M Whittington, D A Parry, J A Vale
ABSTRACT
Isobutyl nitrite is a popular recreational drug among both homosexuals and heterosexuals as it is alleged to enhance sexual pleasure and prolong orgasm. In contrast to the ingestion of this volatile nitrite, inhalation is associated only rarely with serious sequelae, though symptomatic methemoglobinemia may occur. The case reported is the first documented fatality from inhalation of isobutyl nitrite. The widespread use of isobutyl nitrite is a cause for concern and physicians should be aware of the potentially fatal consequence of abuse, particularly in those with ischemic heart disease, and its management.
Sigma-Aldrich
Isobutyl nitrite, 95%