Merck
  • Nidogen-1 is a novel extracellular ligand for the NKp44 activating receptor.
Nidogen-1 is a novel extracellular ligand for the NKp44 activating receptor.
Oncoimmunology (2018-09-20)
Silvia Gaggero, Maurizio Bruschi, Andrea Petretto, Monica Parodi, Genny Del Zotto, Chiara Lavarello, Carola Prato, Laura Santucci, Alessandra Barbuto, Cristina Bottino, Giovanni Candiano, Alessandro Moretta, Massimo Vitale, Lorenzo Moretta, Claudia Cantoni
ABSTRACT
The release of soluble ligands of activating Natural Killer (NK) cell receptors may represent a regulatory mechanism of NK cell function both in physiologic and in pathologic conditions. Here, we identified the extracellular matrix protein Nidogen-1 (NID1) as a ligand of NKp44, an important activating receptor expressed by activated NK cells. When released as soluble molecule, NID1 regulates NK cell function by modulating NKp44-induced IFN-γ production or cytotoxicity. In particular, it also modulates IFN-γ production induced by Platelet-Derived Growth Factor (PDGF)-DD following NKp44 engagement. We also show that NID1 may be present at the cell surface. In this form or when bound to a solid support (bNID1), NID1 fails to induce NK cell cytotoxicity or cytokine release. However, analysis by mass spectrometry revealed that exposure to bNID1 can induce in human NK cells relevant changes in the proteomic profiles suggesting an effect on different biological processes.
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
SKU
Pack Size
Availability
Price
Quantity
SAFC
EX-CELL® ACF CHO Medium, Animal-component free, with HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture