Merck
  • Tartronic acid promotes de novo lipogenesis and inhibits CPT-1β by upregulating acetyl-CoA and malonyl-CoA.
Tartronic acid promotes de novo lipogenesis and inhibits CPT-1β by upregulating acetyl-CoA and malonyl-CoA.
Life sciences (2020-08-12)
Xin'e Shi, Xiaomin Zhou, Jie Wang, Deming Zhang, Kuilong Huang, Xiao Li, Gongshe Yang
ABSTRACT
As a dicarboxylic acid with the structural formula HOOCCH (OH) COOH, tartronic acid is considered as an inhibitor of the transformation of carbohydrates into fat under fat-deficient diet conditions. However, the effect of tartronic acid on lipogenesis under high-fat diet conditions has yet to be established. In this work, we investigated the regulatory role of tartronic acid in lipogenesis in 3T3-L1 adipocytes and C57BL/6J mice. The results confirmed that tartronic acid promoted weight gain (without affecting food intake) and induced adipocyte hypertrophy in epididymal white adipose tissue and lipid accumulation in the livers of high-fat diet-induced obese mice. In vitro, tartronic acid promoted 3T3-L1 adipocyte differentiation by increasing the protein expression of FABP-4, PPARγ and SREBP-1. Moreover, the contents of both acetyl-CoA and malonyl-CoA were significantly upregulated by treatment with tartronic acid, while the protein expression of CPT-1β were inhibited. In summary, we proved that tartronic acid promotes lipogenesis by serving as substrates for fatty acid synthesis and inhibiting CPT-1β, providing a new perspective for the study of tartronic acid.
Sigma-Aldrich
Tartronic acid, ≥97.0%