Merck
  • Total synthesis of 2-hydroxytetracosanolide and 2-hydroxy-24-oxooctacosanolide by using an effective lactonization.
Total synthesis of 2-hydroxytetracosanolide and 2-hydroxy-24-oxooctacosanolide by using an effective lactonization.
Chemistry, an Asian journal (2008-01-19)
Isamu Shiina, Akane Sasaki, Takaaki Kikuchi, Hiroki Fukui
ABSTRACT
We have developed effective methods for the total synthesis of 2-hydroxytetracosanolide and 2-hydroxy-24-oxooctacosanolide, the defensive salivary secretions of termites. The former natural product isolated from Armitermes neotenicus, a species of termite that inhabits Guyana, contains a 25-membered lactone backbone, and the latter, extracted from Pseudacanthoterme springer, an African termite, includes a 29-membered lactone moiety. The key cyclization to produce the 25- or 29-membered lactone core is performed by using 2-methyl-6-nitrobenzoic anhydride (MNBA) with a stoichiometric amount of 4-(dimethylamino)pyridine (DMAP) or a catalytic amount of 4-(dimethylamino)pyridine N-oxide (DMAPO).
Sigma-Aldrich
Benzoic anhydride, ≥95%