Merck
  • [Effects of para-tetra-hydroxyphenyl-porphyrin and red light on kidney clear cell carcinoma cells of Wistar rats].
[Effects of para-tetra-hydroxyphenyl-porphyrin and red light on kidney clear cell carcinoma cells of Wistar rats].
Patologia polska (1990-01-01)
P Ziółkowski, A Małek, L Latos-Grazyński, M Wodnik
ABSTRACT
Results of investigation of para-tetra-hydroxyphenyl-porphyrin and red light on treatment of kidney clear cell carcinoma in Wistar rats have been assessed. Positive influence of photodynamic therapy was confirmed.
Sigma-Aldrich
5,10,15,20-Tetrakis(4-hydroxyphenyl)-21H,23H-porphine, Dye content 95 %