Merck
  • Factors responsible for inhibiting the motility of zoospores of the phytopathogenic fungus Aphanomyces cochlioides isolated from the non-host plant Portulaca oleracea.
Factors responsible for inhibiting the motility of zoospores of the phytopathogenic fungus Aphanomyces cochlioides isolated from the non-host plant Portulaca oleracea.
FEBS letters (1998-11-25)
M Mizutani, Y Hashidoko, S Tahara
ABSTRACT
In a survey of plant secondary metabolites regulating the behaviour of Aphanomyces cochlioides zoospores, we found that root extracts of Portulaca oleracea inhibited zoospore motility. Bioassay-directed fractionation of Portulaca constituents revealed that the inhibitory activity was dependent on the interaction of two chemically different factors. These were identified as a phenolic compound, N-trans-feruloyltyramine, which by itself was active as a zoospore stimulant, and an acidic compound, 1-linoleoyl-2-lysophosphatidic acid monomethyl ester, which had zoospore-repellent activity. When Chromosorb W AW particles coated with a mixture of these pure compounds were bioassayed in Petri dishes, the inhibitory effect on zoospore motility was identical with that caused by root tip or root extracts of P. oleracea. Inhibited zoospores rapidly settled to the bottom of the Petri dishes where they initially encysted, and then germinated within 1-2 h. This is the first report of factors which inhibit zoospore motility without killing or bursting the zoospores.
Sigma-Aldrich
Moupinamide, ≥95% (LC/MS-ELSD)