HomeSmall Molecule HPLCHPLC Analysis of Lactic Acid Enantiomers on Astec® CLC-L, Trace Level Detection
HPLC Analysis of Lactic Acid Enantiomers on Astec® CLC-L, Trace Level Detection
Conditions | |
---|---|
Column | Astec CLC-L, 15 cm x 4.6 mm I.D., 5 µm particles (53123AST) |
Column temp. | 25 °C |
Mobile phase | 5 mM copper sulfate |
Flow rate | 1 mL/min |
Sample | 5 mg/mL in water |
Injection | 10 µL |
Detector | UV, 254 nm |
Description | |
---|---|
Categories | Analytical Chromatography, Chiral Applications, Food & Beverage, Organic Acids, Chemicals and Industrial Polymers, All Food & Beverage Applications |
Featured Industry | Chemicals and Industrial Polymers Food and Beverages Vitamins, Nutraceuticals, and Natural Products |
Legal Information | Astec is a registered trademark of Sigma-Aldrich Co. LLC |
Suitability | Application for HPLC |