Merck
HomeDNA & RNA PurificationProtein Affinity Chromatography
Protein Affinity Chromatography
Affinity chromatography is the preferred method of bioselective adsorption and subsequent recovery of a compound from an immobilized ligand. Each is designed for highly specific and efficient purification of proteins and related compounds. We offer appropriately selective ligands on beaded and porous matrices for binding target compounds, which are then recovered under mild conditions. Your choice for optimal downstream utility.
Affinity ClassPotential Application
Activated / FunctionalizedFunctional spacer; support matrix; eliminates handling of toxic reagents
Amino AcidSerum proteins; proteins; peptides; enzymes; rRNA; dsDNA
Avidin BiotinPurification of biotin/avidin & derivatives; biotinylated substances. Biotin derivatives dissociate under nondenaturing conditions.
Carbohydrate BindingSolube glycoproteins ; other carbohydrate-containing substances
CarbohydrateGlycoproteins; lectins; other carbohydrate metabolite proteins. Proper selection can ensure one-step purification.
Dye LigandNonspecific interaction. Mimic biological substrates (substrates, cofactors, effetors); proteins. Optimize purification protocol with different dyes
GlutathionePurification of glutathione enzymes and GST tagged recombinant proteins
HeparinGeneral affinity ligand, useful for plasma coagulation proteins, nucleic acid enzymes, lipases, etc.
Hydrophobic InteractionsCouple ligands containing free carboxyl groups; proteins
IMACImmobilized Metal Affinity Chromatography. Uses interactions between protein and chelated metal to separate.
ImmunoaffinityQuantitative determinination of antigens, high specificity
Nucleotide / CoenzymeDehydrogenases; kinases; transaminases. Reliable adsorbents.
Nucleic AcidmRNA; DNA; rRNA; other nucleic acids and oligonucleotides
Protein A / Protein GPurification of immunoglobulins
SpecialityPurification of specific classes or types of proteins, coenzymes, or physiological partners
Applications for Affinity Media
Activated / Functionalized Matrices
The availability of pre-activated supports allows the user to quickly and easily custom synthesize affinity resins without having to handle highly reactive reagents. We offer a variety of activated matrices ready for direct coupling to a diversity of ligands. The ligand, generally, must possess a free primary amine, sulfhydryl, or hydroxyl group for direct attachment. Ligand characteristics will be a major determinant in choosing an appropriate matrix.
Historically, cyanogen bromide activated matrices have been very popular. Their ability to efficiently couple amine functionalities under mild conditions has, for many, offset the problems of ligand leaching and ionic properties associated with this chemistry. We offer many alternative activated resins with coupling chemistries suitable for stable attachment to most types of potential ligands. Different activating chemistries will impart unique characteristics to the reactivity and functionality of the affinity matrix. The major properties for each direct activation type are described in the accompanying table.
ActivationLinkage to ResinAvailable Reactive GroupSpecificity of GroupReaction ConditionsBond Type; Stability of AttachmentIntrinsic Spacer
CarbonyldimidazoleCarbamateImidazole CarbamateAminepH 8-10Carbamate; good stability below pH 101 atom neutral
Cyanogen BromideFinal isoureaCyanate esterAminepH 8-9.5Isourea; moderately stable1 atom cationic
EpichlorohydrinEtherEpoxySH>NH2>OHpH 7-8 SHThioether sec amine ether; all very stable3 atoms neutral
Epoxy (bis)EtherEpoxySH>NH2>OHpH 7-8 SH
pH 9-11 NH2
pH >11 OH
Thioether sec amine ether; all very stable12 atoms neutral
N-Hydroxy-succinimidyl ChloroformateFinal carbonateSuccinimidyl CarbonateAminepH 8-9.5Carbamate; good stability below pH 101 atom neutral
p-Nitrophenyl ChloroformateFinal carbonateNitrophenyl CarbonateAminepH 8.5-10Carbomate; good stability below pH 101 atom neutral
Tresyl ChlorideAlkylamine thioetherTresyl sulfonateAmine, thiolpH 7-9.5Alkylamine thioether; both very stable0 atoms direct linkage
Vinyl SulfoneSulfonyl thioetherVinyl sulfoneSH>NH2>OHpH 6-8 SH
pH 8-10 NH2
pH >10 OH
ThiotherSec amine ether; Good stability below pH 95 atoms neutral, some non-specific effects
Direct Activated Matrices
Some activated matrices are available which contain an additional spacer due to prior derivatization. These resins may permit the use of milder coupling conditions or a more specific attachment of a ligand. Examples of these are as follows:
Active GroupSpecificityCoupling ConditionsBond to ligand; stability
N-Hydroxysuccinimide Ester; active esterAminepH 6.0-8.0Amine; good stability
Disulfide; reactivity based on leaving groupSulfhydrylpH 6.0-8.0Covalent disulfide; good stability under nonreducing conditions
Activated Groups Incorporated to Prederivatized Matrices
We offer a variety of matrices with incorporated terminal groups suitable for custom derivatization or coupling. Typically these groups will be a carboxyl or amine function, which may be coupled by an amide bond to a ligand, and require an additional reagent to accomplish condensation. Another group commonly utilized is hydrazide, which may couple to aldehydes by hydrazone formation and typically does not require additional reagent.
Amino Acid Resins
We offer a variety of amino acids coupled through the amino group to cyanogens bromide activated 4% beaded agarose. These resins may be used directly for adsorpotion of compounds having an affinity for the particular amino acid immobilized, or they can serve as functionalised linkers and allow covalent attachment through their free carboxyl terminus to a ligand containing a free primary amine.
Avidin / Biotin Matrices
The protein Avidin has an extremely high affinity for the cofactor Biotin (Kdiss 10-15M). This characteristic provides a unique and powerful tool, which can be used for separation and purification. Many compounds are easily biotinylated with retention of biological activity, allowing for a broad range of separation applications. We offer a wide variety of biotinylated products, biotinylation reagents and avidin derivates.
One disadvantage of the strong interaction between avidin and biotin is the harsh denaturing conditions required to accomplish dissociation. Harsh conditions may be avoided by using a biotin derivative such as 2-iminobiotin, which dissociates from avidin under milder conditions. Another alternative, is the use of monomeric avidin, which has a much lower affinity for biotin than the natural tetramer. This allows for dissociation of the complex by competitive displacement.
Carbohydrate Binding Matrices
Lectins are the proteins that have the ability to bind to sugar moieties and agglutinate cells. Once immobilized, lectins can be utilized for the purification of selected glycoconjugates, which may generally be recovered by competitive displacement using an inhibitory simple sugar. Lectin resins have been used to:
  • Purify polysaccharides
  • Fractionate cell parts
  • Purify glycoproteins and other glycoconjugate molecules
  • Remove carbohydrate containing impurities from a protein solution
Sugar SpecificityLectin
β-D-gal(1-3)-D-galNAcArahis hypogaea (Peanut)
Methyl-α-D-galArtocarpus integrifolia (Jacalin)
α-D-man, α-D-glcConcanavalin A Type VI (Con A)
α-D-man, α-D-glcSuccinyl Concanvalin A
Nonreducing end of terminal α-D-mannosyl residue of glycoconjugatesGalanthus nivalis (Snowdrop)
D-galNAcGlycine max (Soybean)
D-galNAcHelix pomatia (Roman or edible snail)
α-D-manLens culinaris (Lentil)
OligosaccharidePhaseolus vulgaris PHA-E (Red kidney bean)
(D-glcNAc)2, NeuNAcTriticum vulgaris (Wheat germ)
α-L-fucUlex europaeus UEA-1 (gorse of Furze)
Carbohydrate Matrices
Immobilized sugars and sugar derivatives offer an effective means of purification for a variety of proteins, which recognize and bind carbohydrate moieties. Among the most widely used applications for these matrices are the purification of lectins, glycosidases, and carbohydrate directed antibodies. Lectin isolation is of particular interest in that they are highly useful for studies of cell membrane structure and function, determination of blood groups, cell fractionation, and stimulation of lymphocyte mitosis.
Dye Ligand Resins
Dye ligand chromatography is affinity chromatography that utilizes covalently bond textile dye (reactive dyes) to purify proteins. These dyes resemble natural substrates which proteins have affinities for, thus dye ligand chromatography is sometimes referred to as pseudo-affinity chromatography. Immobilized dyes are not specific adsorbents, but rather have a broad binding range for a variety of proteins. After careful investigation of the proper binding and elution conditions, a high degree of purification of a target protein can be attained even in the presence of a mixture of other proteins.
Glutathione Resins
Glutathione is a tri-peptide, which consists of the amino acids Glutamic acid, Cysteine and Glycine. Glutathione and its derivatives are very useful as ligands in affinity chromatography for the isolation of Glutathione requiring enzymes. These include detoxification enzymes such as: Glutathione Transferase, Glutathione Peroxidase and Glyoxalase I. In recent years, Glutathione agarose resins have become useful for the isolation of fusion proteins due to the presence of Glutathione transferase (GST) as part of the fusion protein. By fusing GST to recombinant proteins, Glutathione resins can separate a selected target protein from other components.
Heparin Resins
Heparin is a highly sulphated glycosaminoglycan, which has widespread use as a general affinity ligand. Its high degree of sulfation imparts a strong acidic nature to the molecule, therefore it may bind to many substances by ionic interaction. In addition, heparin contains unique carbohydrate sequences, which act as specific binding sites for some proteins. Immobilized heparin has been used to purify plasma coagulation proteins, nucleic acid enzymes, lipases, and other proteins.
Hydrophobic Interaction Resins
Hydrophobic chromatography is a versatile technique used for the separation and purification of proteins based on their hydrophobicity. Columns are usually run under conditions that favor hydrophobic interaction, such as high ionic strength. This makes hydrophobic chromatography an ideal tool to use following salt precipitation. Our hydrophobic gels are available in three functional variations:
  • Pure hydrophobic resins—these gels contain ligands attached through a stable ether linkage and they contain no charged regions
  • Mixed property resins—these gels contain ligands attached through an amine group and in some cases they will carry a charge
  • Aminoalkyl resins—these gels provide a hydrophobic spacer to which other functional groups may be coupled
 IMAC Matrices
Immobilized Metal Affinity Chromatography (IMAC) is the process of protein separation based on the differential interaction of various proteins with different chelated / insolubilized metals. This interaction depends on:
  • The relative amount and location of certain amino acids within the protein (namely histidine, cysteine, tyrosine, and typtophan)
  • The type of metal utilized
Generally, different transition metals may be chelated to and exchanged within the same resin, however, some chelating groups may demonstrate preferential chelation of certain metals. Commonly utilized metals are: Copper, Zinc, Iron, and Nickel.
MetalApplication
CobaltPurification of HIS-tagged proteins
NickelPurification of HIS-tagged proteins
CopperUsually for purification of HIS-tagged proteins and histidine containing peptides
ZincUsually for the purification of HIS-tagged proteins
IronPurification of Phosphopeptides
GalliumPurification of Phosphopeptides
 Immunoaffinity Matrices
Immobilized antibodies are used as analytical tools for the quantitative determination of antigens, including immunoglobulins and haptens. Immunoprecipitation followed by SDS-PAGE is a technique typically used to determine the quantity and presence of an antigen in a complex protein mixture such as a cell lysate. Cells may be separated according to surface antigens using various immobilized immunochemicals. Antibody-agarose products are also used in a variety of application including immunoadsorption, affinity chromatography and as solid-phase secondary antibodies. Due to this diversity, we offer a wide selection of antibodies and whole sera immobilized onto 4% crosslinked agarose.
 Nucleotide / Coenzyme Resins
Cofactors, coenzymes, and substrates bound to matrices play a major role in protein purification. Since nearly one third of known enzymes require a nucleotide coenzyme, nucleotide bound resins are useful for the purification of many proteins. Protein binding to a nucleotide coenzyme is dependent on spacer arm length and the position of ligand attachment. We offer a wide variety of nucleotide resins with different attachment positions and spacer arms to provide the chemist greater flexibility in affinity fractionations.
 Protein A / Protein G Matrices
Protein A and Protein G are bacterial proteins, which demonstrate specific binding to the Fc (non-antigen binding) portion of many classes of immunoglobulins. Protein A and G affinity matrices have been used primarily for:
  • Affinity purification of immunoglobulin, primarily IgGs
  • Separation of Fc from Fab fragments
Protein A resins have historically been popular for most potential applications, however it has been demonstrated that Protein G resin can enhance and broaden the scope of application. The binding characteristics of the two proteins for various types of immunoglobulins vary and may be used to good advantage. Some of the major differences in binding are:
  • Protein A
    • Broad species reactivity; binds well to IgG from human, rabbit, cow and guinea pig
    • Weak binding to monoclonal antibodies
  • Protein G
    • Broader species reactivity; much stronger binding to IgG from mouse, rat and goat
    • Stronger binding to monoclonal antibodies
Specialty Resins
The following is a sample list of products containing a variety of specialty affinity matrices. Each resin is listed with possible applications and references.
Resin TypeFor the purification of (Applications), [Reference]
Albumin agaroseSteroids, amino acids, bilirubin & heme peptides (For general hydrophobic adsorption & chiral resolution) 1,2
p-Aminobenzamide agaroseSerine proteases, urokinases, plasminogen activators and protease removal 3,4
m-Aminophenylboronic acid agaroseGlycoproteins (This matrix offers high capacity) 5
N(p-Aminophenyl)oxamic acid agaroseNeuraminidase & influenza virus 6
Antithrombin III agaroseHeparin & thrombin 7,8
Aprotinin agaroseSerine proteases, urokinases & protease removal 9
Cholesterol hemisuccinate agaroseHDL, LDL & cholesterol oxidase 10
α-Cobrotoxin agaroseNicotinic acetylcholine (snake toxin) receptors 12
Fetuin agaroseLectins, glycosidases & carbohydrate binding proteins 12,13
Folic acid agaroseFolate binding proteins 14
Forskolin agaroseAdenylate cyclase 15
Gelatin agaroseFibronectin 16,17
Hemoglobin agaroseHemoglobin based O2 carriers 18
L-Histidyldiazobenzyl phophonic acid agarosePhosphatases 19
Histone agaroseProtein kinases 20
Methotrexate agaroseDihydrofolate reductase 21
Pepstatin A agaroseCathepsin D & Rennin 22
Phosphodiesterase 3’-5’ cyclic nucleotide activator agaroseCalmodulin binding proteins 23,24
O-Phospho-L-Tyrosine agarosePhosphatases 19
Poly-L-Lysine agaroseDNA, RNA & T4 phage 25
Polymyxin B agaroseLipopolysaccharides (endotoxins) 26,27
Tartaric acid agaroseProstatic acid phosphatases 30,31
Trypsin inhibitor agaroseProteases or protease removal 32,33
References
1.
Popov D. 1992. Cardiomyocytes express albumin binding proteins. Journal of Molecular and Cellular Cardiology. 24(9):989-1002. https://doi.org/10.1016/0022-2828(92)91865-3
2.
Allenmark S, Bomgren B, Borén H. 1982. Direct resolution of enantiomers by liquid affinity chromatography on albumin-agarose under isocratic conditions. Journal of Chromatography A. 237(3):473-477. https://doi.org/10.1016/s0021-9673(00)97635-0
3.
Winkler, M. E., et al.. 1985. Biotechnology. 3, 990-1000..
4.
Strickland, Strickland. 1983. Biochem. 22, 444. (Abstract).
5.
Bouriotis V, Galpin IJ, Dean P. 1981. Applications of immobilised phenylboronic acids as supports for group-specific ligands in the affinity chromatography of enzymes. Journal of Chromatography A. 210(2):267-278. https://doi.org/10.1016/s0021-9673(00)97837-3
6.
Cuatrecasas P, Illiano G. 1971. Purification of neuraminidases from Vibrio cholerae, Clostridium perfringens and influenza virus by affinity chromatography. Biochemical and Biophysical Research Communications. 44(1):178-184. https://doi.org/10.1016/s0006-291x(71)80175-4
7.
Byrun, Y. et al. J. Biomater.,. Ed.1994. Sci. Polym.. 6, 1-13.
8.
Thunberg L, Bäckström G, Lindahl U. 1982. Further characterization of the antithrombin-binding sequence in heparin. Carbohydrate Research. 100(1):393-410. https://doi.org/10.1016/s0008-6215(00)81050-2
9.
Johnson DA, Travis J. 1976. Rapid purification of human trypsin and chymotrypsin I. Analytical Biochemistry. 72(1-2):573-576. https://doi.org/10.1016/0003-2697(76)90568-6
10.
Wichman A. 1979. Affinity chromatography of human plasma low- and high-density lipoproteins. Elution by selective cleavage of a bond in the spacer. 181(3):691-698. https://doi.org/10.1042/bj1810691
11.
Mosckovitz, R.; Gershoni, J. M. J. Biol. Chem.1988263, 1017. (Abstract).
12.
Haidar M, Seddiki N, Gluckman JC, Gattegno L. 1992. Carbohydrate binding properties of the envelope glycoproteins of human immunodeficiency virus type 1. Glycoconjugate J. 9(6):315-323. https://doi.org/10.1007/bf00731092
13.
MITSAKOS A, HANISCH F. 1989. One-Step Purification of an ?(1-3)-L-Fucosyltransferase from Human Amniotic Fluid by Fetuin-Agarose Affinity Chromatography. Biological Chemistry Hoppe-Seyler. 370(1):239-244. https://doi.org/10.1515/bchm3.1989.370.1.239
14.
Salter DN, Scott KJ, Slade H, Andrews P. 1981. The preparation and properties of folate-binding protein from cow's milk. 193(2):469-476. https://doi.org/10.1042/bj1930469
15.
Pfeuffer, E. et al. . Proc. Natl. Acad. Sci. USA1985,82, 3086. (Abstract).
16.
Ruoslahti, E. et al. J. Biol.. 1979. Chem.,254, 6054-6059. (Abstract).
17.
Habeeb A. 1981. Controlled coupling of mildly reduced proteins to Sepharose gelatin by heterobifunctional reagent. Biochemical and Biophysical Research Communications. 100(3):1154-1166. https://doi.org/10.1016/0006-291x(81)91945-8
18.
Chiancone E, Fronticelli C, Gattoni M, Urbaitis BK, Bucci E. 1992. Immobilized hemoglobin in the purification of hemoglobin-based oxygen carriers. Journal of Chromatography A. 604(1):117-123. https://doi.org/10.1016/0021-9673(92)85536-3
19.
Landt M, Boltz SC, Butler LG. 1978. Alkaline phosphatase: affinity chromatography and inhibition by phosphonic acids. Biochemistry. 17(5):915-919. https://doi.org/10.1021/bi00598a027
20.
Wallis, M. H. et al. . 1980. Biochem.,19, 798. (Abstract).
21.
Cayley PJ, Dunn SMJ, King RW. 1981. Kinetics of substrate, coenzyme, and inhibitor binding to Escherichia coli dihydrofolate reductase. Biochemistry. 20(4):874-879. https://doi.org/10.1021/bi00507a034
22.
Afting EG, Recker ML. 1981. Two-step affinity-chromatographic purification of cathepsin D from pig myometrium with high yield. 197(2):519-522. https://doi.org/10.1042/bj1970519
23.
Niggli V, Zurini M, Carafoli E. 1987. [58] Purification, reconstitution, and molecular characterization of the Ca2+ pump of plasma membranes.791-808. https://doi.org/10.1016/0076-6879(87)39127-x
24.
Dedman, J. R. et al. J. Biol.. Chem.1993. ,268, 23025-23030. (Abstract).
25.
Sundberg L, Höglund S. 1973. Purification of T4 phage by adsorption on polylysine agarose. 37(1):70-73. https://doi.org/10.1016/0014-5793(73)80428-4
26.
MOLVIG J, BAEK L. 1987. Removal of Endotoxin from Culture Media by a Polymyxin B Sepharose Column.. Scand J Immunol. 26(6):611-619. https://doi.org/10.1111/j.1365-3083.1987.tb02296.x
27.
Issekutz AC. 1983. Removal of gram-negative endotoxin from solutions by affinity chromatography. Journal of Immunological Methods. 61(3):275-281. https://doi.org/10.1016/0022-1759(83)90221-1
28.
Piepkorn MW, Lagunoff D, Schmer G. 1980. Binding of heparin to antithrombin III: The use of dansyl and rhodamine labels. Archives of Biochemistry and Biophysics. 205(2):315-322. https://doi.org/10.1016/0003-9861(80)90113-7
29.
WOOTEN MW, VANDENPLAS M, NEL AE. 1987. Rapid purification of protein kinase C from rat brain. A novel method employing protamine-agarose affinity column chromatography. Eur J Biochem. 164(2):461-467. https://doi.org/10.1111/j.1432-1033.1987.tb11079.x
30.
Van Etten RL, Saini MS. 1978. Selective purification of tartrate-inhibitable acid phosphatases: rapid and efficient purification (to homogeneity) of human and canine prostatic acid phosphatases.. 24(9):1525-1530. https://doi.org/10.1093/clinchem/24.9.1525
31.
Lin MF, Lee CL, Li SSL, Chu TM. 1983. Purification and characterization of a new human prostatic acid phosphatase isoenzyme. Biochemistry. 22(5):1055-1062. https://doi.org/10.1021/bi00274a009
32.
Feinstein G, Hofstein R, Koifmann J, Sokolovsky M. 1974. Human Pancreatic Proteolytic Enzymes and Protein Inhibitors. Isolation and Molecular Properties. Eur J Biochem. 43(3):569-581. https://doi.org/10.1111/j.1432-1033.1974.tb03444.x
33.
Peterson LM, Sokolovsky M, Vallee BL. 1976. Purification and crystallization of human carboxypeptidase A. Biochemistry. 15(12):2501-2508. https://doi.org/10.1021/bi00657a001