47789
2,3-Dibromopropionic acid solution
certified reference material, 1000 μg/mL in methyl tert-butyl ether
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Agency
EPA 552.2
product line
TraceCERT®
CofA
current certificate can be downloaded
feature
standard type surrogate
packaging
ampule of 1 mL
concentration
1000 μg/mL in methyl tert-butyl ether
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
storage temp.
2-8°C
InChI
1S/C3H4Br2O2/c4-1-2(5)3(6)7/h2H,1H2,(H,6,7)
InChI key
ZMYAKSMZTVWUJB-UHFFFAOYSA-N
Application
2,3-Dibromopropionic acid may be used as a surrogate standard for the quantification of haloacids in water samples using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-27.4 °F - closed cup
Flash Point(C)
-33.0 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Solid-phase microextraction coupled with gas chromatography?ion trap mass spectrometry for the analysis of haloacetic acids in water
Journal of Chromatography A, 859(2), 159-171 (1999)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service