Skip to Content
Merck
4-Amino-3,5-dibromopyridine 95%

4-Amino-3,5-dibromopyridine

4-Amino-3,5-dibromopyridine 95%
Synonym(s):
3,5-Dibromo-4-pyridinamine
Empirical Formula (Hill Notation):
C5H4Br2N2
CAS Number:
Molecular Weight:
251.91

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing