Skip to Content
Merck
6-Fluoro-3-(4-piperidinyl)benzisoxazole 95%

6-Fluoro-3-(4-piperidinyl)benzisoxazole

6-Fluoro-3-(4-piperidinyl)benzisoxazole 95%
Synonym(s):
4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidine, 6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole, 6-Fluoro-3-(piperidin-4-yl)benzo[d]isoxazole
Empirical Formula (Hill Notation):
C12H13FN2O
CAS Number:
Molecular Weight:
220.24

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing