6-O-Methylguanine (2-Amino-6-Methoxypurine)
Synonym(s):
6-O-Methylguanine, 2-Amino-6-methoxypurine, 6-Methoxyguanine
Empirical Formula (Hill Notation):
C6H7N5O
CAS Number:
Molecular Weight:
165.15
Products
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing