
Cephalexin monohydrate

Empirical Formula (Hill Notation):
C16H17N3O4S · H2O
CAS Number:
Molecular Weight:
365.40
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing