![(+)-Catechin 5-gallate phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/338/153/621d097f-9e27-4d37-8f23-dc9850e6eaa9/640/621d097f-9e27-4d37-8f23-dc9850e6eaa9.png)
(+)-Catechin 5-gallate
![(+)-Catechin 5-gallate phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/338/153/621d097f-9e27-4d37-8f23-dc9850e6eaa9/640/621d097f-9e27-4d37-8f23-dc9850e6eaa9.png)
Synonym(s):
Catechin 5-O-gallate, 3,4,5-Trihydroxybenzoic acid (2R,3S)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-3,7-dihydroxy-2H-1-benzopyran-5-yl ester
Empirical Formula (Hill Notation):
C22H18O10
CAS Number:
Molecular Weight:
442.37
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing