Isovaleraldehyde-DNPH solution
Synonym(s):
Isovaleraldehyde 2,4-dinitrophenylhydrazone solution
CAS Number:
Products
Brand
Product Line
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing