
Levmetamfetamine

Synonym(s):
(−)-Deoxyephedrine, (−)-2-(Methylamino)-1-phenylpropane, (−)-Methamphetamine
Empirical Formula (Hill Notation):
C10H15N
CAS Number:
Molecular Weight:
149.23
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing