
L-Valine-13C5

Linear Formula:
(13CH3)213CH13CH(NH2)13COOH
CAS Number:
Molecular Weight:
122.11
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing