Menthol solution
Empirical Formula (Hill Notation):
C10H20O
Molecular Weight:
156.27
Products
Brand
Formula Weight
Gene Alias
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing