
Pyrrolidone

Synonym(s):
2-Pyrrolidinone, 2-Pyrrolidone, Butyrolactam
Empirical Formula (Hill Notation):
C4H7NO
CAS Number:
Molecular Weight:
85.10
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing