
(Trimethylsilyl)diazomethane solution

Synonym(s):
(Diazomethyl)trimethylsilane
Linear Formula:
(CH3)3SiCHN2
CAS Number:
Molecular Weight:
114.22
Products
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing