Merck
  • Tick extracellular vesicles enable arthropod feeding and promote distinct outcomes of bacterial infection.
Tick extracellular vesicles enable arthropod feeding and promote distinct outcomes of bacterial infection.
Nature communications (2021-06-19)
Adela S Oliva Chávez, Xiaowei Wang, Liron Marnin, Nathan K Archer, Holly L Hammond, Erin E McClure Carroll, Dana K Shaw, Brenden G Tully, Amanda D Buskirk, Shelby L Ford, L Rainer Butler, Preeti Shahi, Kateryna Morozova, Cristina C Clement, Lauren Lawres, Anya J O' Neal, Choukri Ben Mamoun, Kathleen L Mason, Brandi E Hobbs, Glen A Scoles, Eileen M Barry, Daniel E Sonenshine, Utpal Pal, Jesus G Valenzuela, Marcelo B Sztein, Marcela F Pasetti, Michael L Levin, Michail Kotsyfakis, Steven M Jay, Jason F Huntley, Lloyd S Miller, Laura Santambrogio, Joao H F Pedra
ABSTRACT
Extracellular vesicles are thought to facilitate pathogen transmission from arthropods to humans and other animals. Here, we reveal that pathogen spreading from arthropods to the mammalian host is multifaceted. Extracellular vesicles from Ixodes scapularis enable tick feeding and promote infection of the mildly virulent rickettsial agent Anaplasma phagocytophilum through the SNARE proteins Vamp33 and Synaptobrevin 2 and dendritic epidermal T cells. However, extracellular vesicles from the tick Dermacentor andersoni mitigate microbial spreading caused by the lethal pathogen Francisella tularensis. Collectively, we establish that tick extracellular vesicles foster distinct outcomes of bacterial infection and assist in vector feeding by acting on skin immunity. Thus, the biology of arthropods should be taken into consideration when developing strategies to control vector-borne diseases.
Sigma-Aldrich
Anti-Calnexin Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)